Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342010.1 | NP_001320091.1 | 7 - 8 | 693 | 102 | |
NM_001342011.1 | NP_001320092.1 | 5 - 6 | 877 | 102 | ||
GenBank mRNA | AY142529.1 | - | 5 - 6 | 479 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |