Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119227.4 | NP_567854.1 | 1 - 2 | 594 | 85 | |
GenBank mRNA | AY050958.1 | - | 1 - 2 | 289 | 85 | |
AY133774.1 | - | 1 - 2 | 266 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |