Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119227.4 | NP_567854.1 | 7 - 8 | 1371 | 70 | |
GenBank mRNA | AY050958.1 | - | 7 - 8 | 1066 | 70 | |
AY133774.1 | - | 7 - 8 | 1043 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |