Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119232.3 | NP_194814.2 | 3 - 4 | 633 | 84 | |
NM_202914.1 | NP_974643.1 | 3 - 4 | 573 | 84 | ||
GenBank mRNA | AY267331.1 | - | 3 - 4 | 406 | 84 | |
BT021124.1 | - | 3 - 4 | 505 | 84 | ||
BT025258.1 | - | 3 - 4 | 406 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |