Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119241.6 | NP_567862.3 | 2 - 3 | 440 | 81 | |
GenBank mRNA | BT004086.1 | - | 2 - 3 | 298 | 81 | |
BT005110.1 | - | 2 - 3 | 241 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |