Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119243.4 | NP_194824.1 | 4 - 5 | 663 | 103 | |
GenBank mRNA | AY045621.1 | - | 4 - 5 | 588 | 103 | |
AY058078.1 | - | 4 - 5 | 563 | 103 | ||
AY058852.1 | - | 4 - 5 | 593 | 103 | ||
AY079039.1 | - | 4 - 5 | 490 | 103 | ||
AY088002.1 | - | 4 - 5 | 605 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |