Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119243.4 | NP_194824.1 | 7 - 8 | 1127 | 60 | |
GenBank mRNA | AY045621.1 | - | 7 - 8 | 1052 | 60 | |
AY058078.1 | - | 7 - 8 | 1027 | 60 | ||
AY058852.1 | - | 7 - 8 | 1057 | 60 | ||
AY079039.1 | - | 7 - 8 | 954 | 60 | ||
AY088002.1 | - | 7 - 8 | 1069 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |