Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124803.3 | NP_200234.1 | 1 - 2 | 348 | 70 | |
GenBank mRNA | AF175991.1 | - | 1 - 2 | 132 | 70 | |
AY059820.1 | - | 1 - 2 | 220 | 70 | ||
AY081512.1 | - | 1 - 2 | 132 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |