Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345081.1 | NP_001330550.1 | 6 - 7 | 683 | 84 | |
NM_124806.2 | NP_200237.1 | 8 - 9 | 869 | 84 | ||
GenBank mRNA | AJ243822.1 | - | 8 - 9 | 838 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |