Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124811.5 | NP_568807.1 | 2 - 3 | 316 | 110 | |
GenBank mRNA | AK226402.1 | - | 2 - 3 | 124 | 110 | |
AY099716.1 | - | 2 - 3 | 262 | 110 | ||
BT000287.1 | - | 2 - 3 | 112 | 110 | ||
FJ794601.1 | - | 2 - 3 | 214 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |