Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FQR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203609.1 | NP_001190538.1 | 2 - 3 | 280 | 94 | |
NM_124830.6 | NP_200261.1 | 2 - 3 | 401 | 94 | ||
GenBank mRNA | AY120735.1 | - | 2 - 3 | 206 | 94 | |
BT002194.1 | - | 2 - 3 | 117 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |