Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124850.4 | NP_200280.1 | 2 - 3 | 463 | 123 | |
GenBank mRNA | AK229580.1 | - | 2 - 3 | 335 | 123 | |
KJ138638.1 | - | 2 - 3 | 264 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |