Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036999.1 | NP_001032076.1 | 5 - 6 | 514 | 73 | |
NM_001345117.1 | NP_001330104.1 | 3 - 4 | 329 | 73 | ||
NM_124895.3 | NP_200324.1 | 5 - 6 | 647 | 73 | ||
GenBank mRNA | AF124159.1 | - | 5 - 6 | 564 | 73 | |
AK317295.1 | - | 5 - 6 | 511 | 73 | ||
BT029156.1 | - | 5 - 6 | 537 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |