Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203616.2 | NP_001190545.1 | 3 - 4 | 634 | 96 | |
NM_001345123.1 | NP_001332152.1 | 2 - 3 | 902 | 96 | ||
NM_124921.5 | NP_200350.2 | 3 - 4 | 689 | 96 | ||
GenBank mRNA | KF112069.1 | - | 2 - 3 | 511 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |