Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124932.4 | NP_568825.1 | 1 - 2 | 957 | 74 | |
GenBank mRNA | AF411785.1 | - | 1 - 2 | 888 | 74 | |
AJ272121.1 | - | 1 - 2 | 696 | 74 | ||
AJ277603.1 | - | 1 - 2 | 830 | 74 | ||
AY143907.1 | - | 1 - 2 | 696 | 74 | ||
KJ138637.1 | - | 1 - 2 | 696 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |