Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124932.4 | NP_568825.1 | 2 - 3 | 1109 | 72 | |
GenBank mRNA | AF411785.1 | - | 2 - 3 | 1040 | 72 | |
AJ272121.1 | - | 2 - 3 | 848 | 72 | ||
AJ277603.1 | - | 2 - 3 | 982 | 72 | ||
AY143907.1 | - | 2 - 3 | 848 | 72 | ||
KJ138637.1 | - | 2 - 3 | 848 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |