Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345146.1 | NP_001331632.1 | 3 - 4 | 1015 | 149 | |
NM_124941.3 | NP_200370.1 | 3 - 4 | 1015 | 149 | ||
GenBank mRNA | DQ979876.1 | - | 3 - 4 | 758 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |