Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345148.1 | NP_001318804.1 | 2 - 3 | 480 | 75 | |
NM_124952.5 | NP_568829.2 | 2 - 3 | 485 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |