Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D6PK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345171.1 | NP_001332267.1 | 2 - 3 | 1299 | 96 | |
NM_124973.5 | NP_200402.1 | 2 - 3 | 1053 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |