Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124995.5 | NP_200424.1 | 10 - 11 | 948 | 77 | |
GenBank mRNA | AY093120.1 | - | 10 - 11 | 934 | 77 | |
BT000174.1 | - | 10 - 11 | 914 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |