Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124995.5 | NP_200424.1 | 6 - 7 | 550 | 101 | |
GenBank mRNA | AY093120.1 | - | 6 - 7 | 536 | 101 | |
BT000174.1 | - | 6 - 7 | 516 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |