Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G19110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118030.3 | NP_567574.1 | 4 - 5 | 1022 | 125 | |
NM_179076.1 | NP_849407.1 | 4 - 5 | 992 | 125 | ||
GenBank mRNA | AF360190.1 | - | 4 - 5 | 966 | 125 | |
AY040004.1 | - | 1 - 2 | 84 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |