Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341325.1 | NP_001329718.1 | 2 - 3 | 549 | 95 | |
NM_118031.4 | NP_567575.1 | 2 - 3 | 617 | 95 | ||
NM_179077.2 | NP_849408.1 | 1 - 2 | 1221 | 95 | ||
GenBank mRNA | AB039927.1 | - | 2 - 3 | 473 | 95 | |
AK317548.1 | - | 1 - 2 | 323 | 95 | ||
AK317670.1 | - | 2 - 3 | 485 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |