Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118069.5 | NP_193684.4 | 2 - 3 | 1214 | 79 | |
NM_179078.3 | NP_849409.2 | 2 - 3 | 1214 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |