Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342225.1 | NP_001328575.1 | 1 - 2 | 521 | 107 | |
NM_119519.4 | NP_567929.1 | 1 - 2 | 671 | 107 | ||
NM_179157.3 | NP_849488.1 | 1 - 2 | 671 | 107 | ||
GenBank mRNA | AY059742.1 | - | 1 - 2 | 460 | 107 | |
AY117173.1 | - | 1 - 2 | 403 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |