Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119799.4 | NP_568001.1 | 5 - 6 | 869 | 89 | |
NM_179175.2 | NP_849506.1 | 5 - 6 | 664 | 89 | ||
GenBank mRNA | AJ270299.1 | - | 5 - 6 | 586 | 89 | |
AY056285.1 | - | 5 - 6 | 645 | 89 | ||
AY091432.1 | - | 5 - 6 | 546 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |