Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATU2AF65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204008.1 | NP_001190937.1 | 3 - 4 | 591 | 63 | |
NM_119833.4 | NP_195387.1 | 3 - 4 | 624 | 63 | ||
NM_179178.3 | NP_849509.1 | 3 - 4 | 618 | 63 | ||
NM_202966.4 | NP_974695.1 | 3 - 4 | 618 | 63 | ||
GenBank mRNA | AF462805.1 | - | 3 - 4 | 618 | 63 | |
AY080711.1 | - | 3 - 4 | 617 | 63 | ||
AY143980.1 | - | 3 - 4 | 505 | 63 | ||
BT000965.1 | - | 3 - 4 | 505 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |