Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085037.2 | NP_001078506.1 | 3 - 4 | 526 | 129 | |
NM_119870.2 | NP_568015.1 | 3 - 4 | 559 | 129 | ||
NM_179180.3 | NP_849511.1 | 3 - 4 | 636 | 129 | ||
NM_179181.3 | NP_849512.3 | 3 - 4 | 559 | 129 | ||
GenBank mRNA | BT029750.1 | - | 3 - 4 | 508 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |