Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204022.1 | NP_001190951.1 | 17 - 18 | 1271 | 103 | |
NM_119986.3 | NP_195537.2 | 17 - 18 | 1328 | 103 | ||
NM_179186.1 | NP_849517.1 | 17 - 18 | 1328 | 103 | ||
GenBank mRNA | AJ243198.1 | - | 17 - 18 | 1242 | 103 | |
AY099838.1 | - | 17 - 18 | 1299 | 103 | ||
BT000334.1 | - | 17 - 18 | 1212 | 103 | ||
KJ138715.1 | - | 17 - 18 | 1212 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |