Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119608.3 | NP_195169.3 | 2 - 3 | 1130 | 66 | |
NM_179232.1 | NP_849563.1 | 2 - 3 | 1132 | 66 | ||
NM_179233.2 | NP_849564.1 | 2 - 3 | 1145 | 66 | ||
NM_202953.1 | NP_974682.1 | 2 - 3 | 1132 | 66 | ||
GenBank mRNA | AY074306.1 | - | 2 - 3 | 1132 | 66 | |
AY096454.1 | - | 2 - 3 | 983 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |