Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160815.2 | NP_001154287.1 | 4 - 5 | 2121 | 84 | |
NM_001203993.1 | NP_001190922.1 | 5 - 6 | 1662 | 84 | ||
NM_001203994.1 | NP_001190923.1 | 4 - 5 | 1781 | 84 | ||
NM_001342339.1 | NP_001328125.1 | 4 - 5 | 1893 | 84 | ||
NM_001342340.1 | NP_001328126.1 | 4 - 5 | 1829 | 84 | ||
NM_001342341.1 | NP_001328129.1 | 4 - 5 | 2036 | 84 | ||
NM_001342342.1 | NP_001328130.1 | 3 - 4 | 2327 | 84 | ||
NM_001342343.1 | NP_001328127.1 | 4 - 5 | 2061 | 84 | ||
NM_001342344.1 | NP_001328128.1 | 5 - 6 | 1999 | 84 | ||
NM_119696.5 | NP_195256.3 | 5 - 6 | 1864 | 84 | ||
NM_179234.2 | NP_849565.1 | 4 - 5 | 1863 | 84 | ||
GenBank mRNA | AJ532570.1 | - | 4 - 5 | 1557 | 84 | |
AY094465.1 | - | 4 - 5 | 1795 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |