Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100191.4 | NP_563674.1 | 3 - 4 | 644 | 154 | |
NM_179252.3 | NP_849583.1 | 3 - 4 | 778 | 154 | ||
GenBank mRNA | AY070723.1 | - | 3 - 4 | 635 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |