Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100365.4 | NP_563720.1 | 5 - 6 | 1108 | 79 | |
NM_179260.1 | NP_849591.1 | 5 - 6 | 1019 | 79 | ||
GenBank mRNA | AY058846.1 | - | 5 - 6 | 988 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |