Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123759.1 | NP_001117231.1 | 5 - 6 | 1069 | 84 | |
NM_001197982.1 | NP_001184911.1 | 7 - 8 | 1364 | 84 | ||
NM_001331528.1 | NP_001322680.1 | 6 - 7 | 1354 | 84 | ||
NM_001331529.1 | NP_001322678.1 | 5 - 6 | 1221 | 84 | ||
NM_001331530.1 | NP_001322679.1 | 6 - 7 | 1267 | 84 | ||
NM_001331531.1 | NP_001322673.1 | 5 - 6 | 2441 | 84 | ||
NM_001331532.1 | NP_001322674.1 | 6 - 7 | 2350 | 84 | ||
NM_001331533.1 | NP_001322675.1 | 6 - 7 | 2323 | 84 | ||
NM_001331534.1 | NP_001322676.1 | 4 - 5 | 2028 | 84 | ||
NM_001331535.1 | NP_001322677.1 | 5 - 6 | 1937 | 84 | ||
NM_100402.3 | NP_172015.1 | 5 - 6 | 1910 | 84 | ||
NM_179265.2 | NP_849596.1 | 6 - 7 | 1373 | 84 | ||
GenBank mRNA | AK316776.1 | - | 6 - 7 | 1278 | 84 | |
AY037177.1 | - | 5 - 6 | 1067 | 84 | ||
AY091690.1 | - | 4 - 5 | 963 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |