Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123759.1 | NP_001117231.1 | 8 - 9 | 1544 | 65 | |
NM_001197982.1 | NP_001184911.1 | 10 - 11 | 1839 | 65 | ||
NM_001331528.1 | NP_001322680.1 | 9 - 10 | 1829 | 65 | ||
NM_001331529.1 | NP_001322678.1 | 8 - 9 | 1696 | 65 | ||
NM_001331530.1 | NP_001322679.1 | 9 - 10 | 1742 | 65 | ||
NM_001331531.1 | NP_001322673.1 | 8 - 9 | 2916 | 65 | ||
NM_001331532.1 | NP_001322674.1 | 9 - 10 | 2825 | 65 | ||
NM_001331533.1 | NP_001322675.1 | 9 - 10 | 2798 | 65 | ||
NM_001331534.1 | NP_001322676.1 | 7 - 8 | 2503 | 65 | ||
NM_001331535.1 | NP_001322677.1 | 8 - 9 | 2412 | 65 | ||
NM_100402.3 | NP_172015.1 | 8 - 9 | 2385 | 65 | ||
NM_179265.2 | NP_849596.1 | 9 - 10 | 1848 | 65 | ||
GenBank mRNA | AK316776.1 | - | 9 - 10 | 1753 | 65 | |
AY037177.1 | - | 8 - 9 | 1542 | 65 | ||
AY091690.1 | - | 7 - 8 | 1438 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |