Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100484.4 | NP_172094.1 | 1 - 2 | 503 | 118 | |
NM_179267.2 | NP_849598.1 | 1 - 2 | 410 | 118 | ||
GenBank mRNA | AY045794.1 | - | 1 - 2 | 369 | 118 | |
AY079377.1 | - | 1 - 2 | 179 | 118 | ||
X95572.1 | - | 300 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |