Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331755.1 | NP_001322007.1 | 12 - 13 | 5205 | 69 | |
NM_100680.4 | NP_563806.1 | 12 - 13 | 5202 | 69 | ||
NM_179277.1 | NP_849608.1 | 11 - 12 | 4934 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |