Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331755.1 | NP_001322007.1 | 10 - 11 | 4890 | 86 | |
NM_100680.4 | NP_563806.1 | 10 - 11 | 4887 | 86 | ||
NM_179277.1 | NP_849608.1 | 9 - 10 | 4619 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |