Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035934.2 | NP_001031011.1 | 8 - 9 | 1202 | 125 | |
NM_100797.6 | NP_172398.1 | 8 - 9 | 1289 | 125 | ||
NM_179292.3 | NP_849623.1 | 8 - 9 | 1289 | 125 | ||
GenBank mRNA | AK316689.1 | - | 8 - 9 | 1186 | 125 | |
AY090944.1 | - | 8 - 9 | 1212 | 125 | ||
AY117313.1 | - | 8 - 9 | 1114 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |