Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084115.2 | NP_001077584.1 | 2 - 3 | 1705 | 100 | |
NM_001332585.1 | NP_001319066.1 | 2 - 3 | 1680 | 100 | ||
NM_102156.1 | NP_564189.1 | 2 - 3 | 1625 | 100 | ||
NM_179369.2 | NP_849700.1 | 2 - 3 | 1717 | 100 | ||
GenBank mRNA | AF087820.1 | - | 2 - 3 | 1511 | 100 | |
AK316806.1 | - | 2 - 3 | 1535 | 100 | ||
AY052356.1 | - | 2 - 3 | 1628 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |