Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA-ADAPTIN 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332624.1 | NP_001322140.1 | 10 - 11 | 1983 | 81 | |
NM_102238.2 | NP_173802.1 | 11 - 12 | 1563 | 81 | ||
NM_179370.4 | NP_849701.1 | 10 - 11 | 1720 | 81 | ||
GenBank mRNA | BT004307.1 | - | 10 - 11 | 1636 | 81 | |
BT005557.1 | - | 10 - 11 | 1420 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |