Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332806.1 | NP_001321133.1 | 6 - 7 | 420 | 90 | |
NM_102586.3 | NP_174142.1 | 6 - 7 | 420 | 90 | ||
NM_179388.2 | NP_849719.1 | 6 - 7 | 420 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |