Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160906.1 | NP_001154378.1 | 1 - 2 | 159 | 76 | |
NM_001332808.1 | NP_001319096.1 | 1 - 2 | 333 | 76 | ||
NM_001332809.1 | NP_001320602.1 | 1 - 2 | 283 | 76 | ||
NM_102599.3 | NP_564305.1 | 1 - 2 | 162 | 76 | ||
NM_179389.4 | NP_849720.1 | 1 - 2 | 159 | 76 | ||
NM_179390.1 | NP_849721.1 | 1 - 2 | 157 | 76 | ||
GenBank mRNA | AF053746.1 | - | 1 - 2 | 142 | 76 | |
AK317149.1 | - | 1 - 2 | 157 | 76 | ||
AY037227.1 | - | 1 - 2 | 158 | 76 | ||
AY074825.1 | - | 1 - 2 | 109 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |