Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332851.1 | NP_001320501.1 | 2 - 3 | 647 | 105 | |
NM_102680.2 | NP_174233.2 | 2 - 3 | 255 | 105 | ||
NM_179396.3 | NP_849727.1 | 2 - 3 | 404 | 105 | ||
GenBank mRNA | AY062613.1 | - | 2 - 3 | 291 | 105 | |
AY070368.1 | - | 2 - 3 | 294 | 105 | ||
AY091327.1 | - | 1 - 2 | 131 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |