Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103882.4 | NP_564559.1 | 6 - 7 | 1100 | 87 | |
NM_179458.3 | NP_849789.1 | 6 - 7 | 1113 | 87 | ||
NM_202269.2 | NP_973998.1 | 6 - 7 | 1189 | 87 | ||
GenBank mRNA | AY062736.1 | - | 6 - 7 | 1025 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |