Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179475.1 | NP_849806.1 | 6 - 7 | 602 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |