Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179491.3 | NP_849822.1 | 3 - 4 | 810 | 82 | |
GenBank mRNA | AF367304.1 | - | 3 - 4 | 715 | 82 | |
BT046174.1 | - | 3 - 4 | 446 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |