Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112103.3 | NP_187874.2 | 11 - 12 | 1565 | 80 | |
GenBank mRNA | AY024357.1 | - | 11 - 12 | 1565 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |