Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112110.4 | NP_566435.1 | 4 - 5 | 772 | 112 | |
GenBank mRNA | AF410309.1 | - | 4 - 5 | 401 | 112 | |
AY045904.1 | - | 4 - 5 | 477 | 112 | ||
AY087607.1 | - | 4 - 5 | 566 | 112 | ||
AY091210.1 | - | 4 - 5 | 342 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |