Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112125.3 | NP_566437.1 | 2 - 3 | 500 | 69 | |
GenBank mRNA | AY085594.1 | - | 2 - 3 | 502 | 69 | |
AY885244.1 | - | 2 - 3 | 476 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |